Learn More
Mineral Oil Rotational Viscosity Standard, Paragon Scientific™
Product Code. 13536020
Change view
Click to view available options
Viscosity:
101.4 mPa.s (25°C)
150.0 mPa.s (25°C)
75.19 mPa.s (25°C)
Unit Size:
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity unitSize
13536020 75.19 mPa.s (25°C) 500mL
13526020 101.4 mPa.s (25°C) 500mL
13546020 150.0 mPa.s (25°C) 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13536020 Supplier Paragon Scientific Supplier No. RTM13
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Paragon ISO 17025 / ISO Guide 34 certified mineral oil rotational viscosity standards are specifically formulated for use with rotational viscometers.
  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in strict accordance with ASTM D2162 at 20°C and 25°C
  • Density g/mL in accordance with ASTM D1480 at 20°C and 25°C
  • Kinematic viscosity at other temperatures are calculated in accordance with ASTM D341
  • Densities at other temperatures are derived from proportional calculations of the measurements at 20°C and 25°C
  • 500mL inert glass packaging with tamper evident cap provides absolute assurance of sample integrity
  • Full traceability to National Standards
  • 12 month shelf life
  • Manufactured in the United Kingdom
Specifications
Viscosity 75.19 mPa.s (25°C)
Certifications/Compliance ISO 17025, UKAS ISO 17034
Quantity 500 mL
Nominal values quoted