Learn More
Mini-Extractor Hoods
Product Code. 11798004
Change view
Click to view available options
Depth (Metric):
300 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
11798004 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11798004 Supplier FTM technologies Supplier No. HI8V
  • ONLINE EXCLUSIVE PRICE
    617.00€
    505.28€ / Pack of 1
    Save 111.72€
    18% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.