missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mini Gel Tank Lid

Product Code. 15394564
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15394564 1 unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15394564 Supplier Invitrogen™ Supplier No. A25944

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Mini Gel Tank Lid is a replacement part for the Mini Gel Tank. It goes on top of the Gel Runner Tank.

About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gels
Gel Compatibility Novex™ Mini Gels
Gel Size Mini
Type Mini Gel Tank Lid
Quantity 1 unit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.