Learn More
Corning™ Mini Microcentrifuge
Compact, lightweight, and easy-to-use
Brand:  Corning™ 6766
Additional Details : Weight : 1.26000kg
 View more versions of this product
Product Code. 15693894
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
The Corning LSE mini microcentrifuge is a personal benchtop instrument designed for quick spin downs of micro-samples. Operation is simple and convenient. After loading sample tubes, close the lid and the rotor accelerates quickly to 6,000 rpm (2,000 x g). This speed range is ideal for bringing small droplets to the bottom of the tubes, micro-filtrations, or basic separations. Press the lid release button and rotor comes to a stop.
The LSE mini microcentrifuge includes an 8-place rotor that will accept standard 1.5 mL to 2.0 mL microcentrifuge tubes, and adapters are included for 0.5/0.4 mL and 0.2 mL tubes. Also included is a 4-place rotor for PCR strip tubes.
  • Compact, lightweight, and easy-to-use
  • Close the lid to start, press the lid release button to stop
  • 8 x 1.5/2.0 mL rotor and 4-place strip tube rotors included
  • Fixed speed of 6,000 rpm generates 2,000 x g.
Specifications
Specifications
Microcentrifuge
8-place rotor that will accept standard 1.5mL to 2.0mL microcentrifuge tubes, and adapters are included for 0.5/0.4mL and 0.2mL tubes. Also included is a 4-place rotor for PCR strip tubes.
6766