Learn More
Mecmesin™ Miniature Button Load Cell
Product Code. 11943978
Encompass
Change view
Click to view available options
Capacity (Metric):
1000 N
2500 N
5000 N
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11943978 5000 N Each
11923978 1000 N Each
11933978 2500 N Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11943978 Supplier Mecmesin™ Supplier No. 878013
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Used for compression only, and is ideal for applications in restricted spaces.
  • Use the S type load cells where space is not limited; dedicated fixtures attach via threaded holes. Static torque screwdrivers may be handheld or mounted.
Specifications
Capacity (Metric) 5000 N