Learn More
Mecmesin™ Miniature Button Load Cell
Used for compression only, and is ideal for applications in restricted spaces.
1876.50€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11923978
|
Mecmesin™
878011 |
1000 N |
1876.50€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11933978
|
Mecmesin™
878012 |
2500 N |
1876.50€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11943978
|
Mecmesin™
878013 |
5000 N |
1876.50€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Use the “S” type load cells where space is not limited; dedicated fixtures attach via threaded holes. Static torque screwdrivers may be handheld or mounted.