missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Minimum Essential Medium Eagle (modified) with Earle's Salts
Shop All MP Biomedicals ProductsDescription
- Cell culture media.
- MP Bio MEM with Earle's salts.
- MEM, originally prepared by Harry Eagle, is one of the most popular cell culture media. Upon his attempts to cultivate normal mammalian fibroblasts and certain HeLa cell subtypes, it was revealed that the nutritional needs of these cell types could not be met by BME. Further studies led to the development of MEM incorporating specific modifications such as higher amino acid concentrations for the cultivation of fastidious cells.
- MP's MEM may be used to support the growth of cells in monolayers, in suspension and wide variety of other cell types with proper supplementation.
Specifications
Specifications
| Form | Solution |
| Product Type | Cell Culture Media |
| Sterility | Non-sterile |
| Quantity | 10 L |
| Osmolality | 295 mOsm/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?