missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIPEP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11002-1-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.Specifications
| MIPEP | |
| Polyclonal | |
| Unconjugated | |
| MIPEP | |
| HMIP, MIP, MIPEP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4285 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99797 | |
| MIPEP | |
| MIPEP Fusion Protein Ag1451 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction