missing translation for 'onlineSavingsMsg'
Learn More

MISA Standard 1: Ge, Hf, Mo, Nb, Si, Ta, Ti, W, Zr @ 100 μg/mL in H2O/tr. HF, VHG™

Product Code. 17972016
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972016 100 mL 100mL
17971336 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17972016 Supplier VHG Labs Supplier No. VHGMISA1100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

MISA Standard 1: Ge, Hf, Mo, Nb, Si, Ta, Ti, W, Zr @ 100 μg/mL in H2O/tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungsten/Germanium/Hafnium/Molybdenum/Niobium/Silicon/Tantalum/Titanium/Zirconium
CAS 7440-33-7, 7440-56-4, 7440-58-6, 7439-98-7, 3-1-7660, 7440-21-3
Methods ICP/ICP-MS
Analyte or Component Names Tungsten/Germanium/Hafnium/Molybdenum/Niobium/Silicon/Tantalum/Titanium/Zirconium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Water
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Tungsten 99.83 μg/mL/Germanium 100 μg/mL/Hafnium 100 μg/mL/Molybdenum 100.1 μg/mL/Niobium 100.1 μg/mL/Silicon 100.2 μg/mL/Tantalum 100.2 μg/mL/Titanium 100.2 μg/mL/Zirconium 100.2 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.