Learn More
MITD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17264-1-AP-150UL
This item is not returnable.
View return policy
Description
MITD1 may play a role in endosomal protein transport.Specifications
| MITD1 | |
| Polyclonal | |
| Unconjugated | |
| MITD1 | |
| MITD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 129531, 363219, 69028 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5I0J5, Q8VDV8, Q8WV92 | |
| Mitd1 | |
| MITD1 Fusion Protein Ag10998 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |