missing translation for 'onlineSavingsMsg'
Learn More

Mithramycin A, TRC

Product Code. 30483559
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483559 1 mg 1mg
30482770 25 mg 25mg
30472404 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483559 Supplier Toronto Research Chemicals Supplier No. TRCM3695001MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18378-89-7
Molecular Formula C52H76O24
Molecular Weight (g/mol) 1085.15
Synonym [1S(2S,3S)]-5-Deoxy-1-C-[7-[[2,6-dideoxy-3-O-(2,6-dideoxy-β-D-arabino-hexopyranosyl)-β-D-arabino-hexopyranosyl]oxy]-3-[(O-2,6-dideoxy-3-C-methyl-β-D-ribo-hexopyranosyl-(1→3)-O-2,6-dideoxy-β-D-arabino-hexopyranosyl-(1→3)-2,6-dideoxy-β-D-lyxo-hexopyranosyl)oxy]-1,2,3,4-tetrahydro-5,10-dihydroxy-6-methyl-4-oxo-2-anthracenyl]-1-O-methyl-D-threo-2-pentulose, Mithramycin, A 2371, Antibiotic LA 7017, Aureolic Acid, Mithracin, Mitramycin, NSC 24559, PA 144, Plicamycin, Plicatomycin
SMILES CC(=O)NCCc1c2c(ccc(O[C@H]3C(OC(C)=O)C(OC(C)=O)[C@@H](OC(C)=O)C(C(=O)OC)O3)c2)n(C(=O)OC(C)(C)C)c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C52H76O24/c1-18-29(72-34-14-30(43(58)21(4)68-34)73-33-13-28(54)42(57)20(3)67-33)12-26-10-25-11-27(49(66-9)48(63)41(56)19(2)53)50(47(62)39(25)46(61)38(26)40(18)55)76-36-16-31(44(59)23(6)70-36)74-35-15-32(45(60)22(5)69-35)75-37-17-52(8,65)51(64)24(7)71-37/h10,12,19-24,27-28,30-37,41-45,49-51,53-61,64-65H,11,13-17H2,1-9H3/t19-,20-,21-,22-,23-,24-,27+,28-,30-,31-,32-,33+,34+,35+,36+,37+,41+,42-,43-,44-,45+,49+,50+,51-,52+/m1/s1
Chemical Name or Material Mithramycin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.