missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitoferrin 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26469-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.Specifications
| Mitoferrin 1 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A37 | |
| MFRN, Mfrn1, Mitochondrial iron transporter 1, Mitoferrin 1, SLC25A37 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51312, 67712 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q920G8, Q9NYZ2 | |
| SLC25A37 | |
| Mitoferrin 1 Fusion Protein Ag24876 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction