missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, MP Biomedicals™

Product Code. 17670867 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 mg
10 mg
20 mg
Unit Size:
10mg
20mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17670867 2 mg 2mg
17680867 10 mg 10mg
17690867 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 17670867 Supplier MP Biomedicals Supplier No. 0210049802
  • ONLINE EXCLUSIVE PRICE
    136.00€
    129.15€ / 2mg
    Save 6.85€
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

The most widely used Mitomycin antibiotic. Mitomycin C, MP Biomedicals is the least toxic of the mitomycin antibiotics, with the most potent antitumor activity. It is an inhibitor of DNA synthesis, nuclear division, and cancer cells.

  • Antibacterial to gram positive, gram negative and acid-fast bacilli
  • Decomposes DNA in cancer cells by forming a complex with DNA
  • Inhibits division of cancer cells by interfering with the biosynthesis of DNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym Ametycin, 7-Amino-9-a-methoxymitosane, Mit-C, Mitocin-C, Mitomycinum, Mutamycin, Azirizino[2', 3': 3,4] pyrrolo [1,2-a] indole 4,7-dione-6-amino-1,1a, 2, 8, 8a, 8b-hexahydro- 8- (hydroxymethyl) 8a-methoxy-5-methyl carbamate
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Physical Form Powder
pH 6 to 9 (Stability)
Extinction Coefficient EmM(367 nm) = 21.84 (water); EmM(357 nm) = 22.78 (methanol); E1% (216 nm) = 742 (methanol); E1% (360 nm) = 742 (methanol); E1% (560 nm) = 0.06 (methanol) (Lit.)
Potency 850μg/mg minimum
Quantity 2 mg
Solubility Information Soluble in dilute aqueous acids or dilute aqueous bases.
Formula Weight 334.33
Grade Antibiotic
compliance-icons compliance-icons
Product Identifier
  • Mitomycin C, Antibiotic Class
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Mitomycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.