missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Mitomycin ReadyMade™ Solution

Product Code. 30044043
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044043 1 mL 1mL
30044014 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044043 Supplier TOKU-E™ Supplier No. M1221ML
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mitomycin ReadyMade™ Solution is provided as a sterile-filtered solution of Mitomycin C in DMSO.

  • CAS Number: 50-07-7
  • Molecular Formula: C15H18N4O5
  • Molecular Weight: 334.33 g/mol
  • Storage Conditions: -20°C, protect from light
  • Form: Liquid
  • Source: Streptomyces caespitosus
  • Spectrum: Mitomycin C is broad-spectrum, effective for Gram-negative and Gram-positive bacteria
  • Mechanism of Action: Mitomycin C is generally non-reactive in its natural oxidized state, and is activated in vivo via reduction, generating oxygen radicals which produce crosslinks in DNA by alkylation. The Mitomycin C itself does not react with DNA. Rather, upon reduction of the quinone, a transformation ensues, and the aziridine ring opens to produce the unstable vinylogous quinone methide 2, which has high alkylating reactivity. High content of guanine and cytosine favor this cross-linking reaction, which is the basis of its lethal effect in vivo

Specifications

CAS 50-07-7
Chemical Name or Material Mitomycin C
Color Purple
Content And Storage -20°C, Protect from light
Description Mitomycin ReadyMade™ Solution 10 mg/mL in DMSO
Concentration 10.0 to 10.5 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in DMSO
For Use With (Application) Eukaryotic Cell Culture, Cancer Application
Product Line ReadyMade™ Solutions
Quantity 1 mL
Shipping Condition Ice Pack
Source Streptomyces caespitosus
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.