Learn More
behr Labor-Technik™ Mixed-Bed Ion Exchanger Made Of Stainless Steel
Product Code. 10799351
Change view
Click to view available options
Flow Rate:
500 L/hr.
700 L/hr.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10799351 500 L/hr. Each
10598092 700 L/hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10799351 Supplier behr Labor-Technik™ Supplier No. B00223482
  • ONLINE EXCLUSIVE PRICE
    2901.86€
    1230.84€ / Pack of 1
    Save 1671.02€
    57% Off
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Standard cartridge made of V4A stainless steel for general purpose application: To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
  • Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
  • Flow rates up to 700 l/h
  • The unique behropur™ jet nozzle system distributes the raw water over the entire resin bed. This ensures an optimum exchange capacity and quality
  • Can be connected to the raw water network directly and without a pressure reducer
  • Resistant to alternating pressure
  • Hard rubber collars vulcanized to the base and top of the unit offer effective protection
  • Convenient transport thanks to the handles embedded into the top part of the unit
  • Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
  • Also available with shutoff when the limited value is reached and limit control
  • Specifications
    Height (Metric) 74 mm
    Type E40d Pressure resistant mixed-bed unit made of V 4 A special alloy, permanent pressure resistant to 10 bar, complete with conductivity meter
    Flow Rate 500 L/hr.