Learn More
Cytiva Mixer
Product Code. 15249774 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15249774 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15249774 Supplier Cytiva Supplier No. 29023960
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use in protein purification applications
The AKTA start mixer is a 0.4mL static mixer used for blending buffers during gradient runs, ensuring reproducibility across protein purification runs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) ÄKTA Start, Blending Buffers During Gradient Runs, Ensuring Reproducibility Across Purification Runs
Quantity 1 Each