missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixture of S077210 (150mL of 1mg/mL S077210 in a 0.9% (Sodium Chloride) NaCl solution), TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Hexachloroplatinate(IV) Hexahydrate |
| CAS | 19583-77-8 |
| Molecular Formula | Cl6Pt • 2Na • 6H2O |
| Quantity | 150 mL |
| Synonym | Platinate(2-), hexachloro-, disodium, hexahydrate |
| InChI Formula | InChI=1S/6ClH.2Na.6H2O.Pt/h6*1H;;;6*1H2;/q;;;;;;2*+1;;;;;;;+4/p-6 |
| SMILES | [Cl-][Pt+4]([Cl-])([Cl-])([Cl-])([Cl-])[Cl-].[Na+].O.O.O.O.O.[Na+].O |
| IUPAC Name | sodium hexachloroplatinate(IV) hexahydrate |
| Molecular Weight (g/mol) | 561.88 |
| Formula Weight | 558.82 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?