missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mkl1 Antibody (CL4281) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59047JF669
This item is not returnable.
View return policy
Description
Mkl1 Monoclonal antibody specifically detects Mkl1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Mkl1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| AMKL, KIAA1438, MALSAP and coiled-coil domain, megakaryoblastic leukemia (translocation) 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, MKL/myocardin-like protein 1, Myocardin-related transcription factor A, RNA-binding motif protein 15/megakaryoblastic leukemia-1 fusion protein | |
| This antibody was developed using a recombinant protein derived from Q969V6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Signal Transduction, Stem Cells | |
| 57591 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4281 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction