missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLC2v Mouse, Clone: S53-5, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565497
This item is not returnable.
View return policy
Description
MLC2v Monoclonal antibody specifically detects MLC2v in Human, Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| MLC2v | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| S53-5 | |
| Unconjugated | |
| MLC-2; MLC-2v; MYL2; MLC2; CMH10 | |
| Human MLC2v | |
| RUO | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction