Learn More
MLH1 Monoclonal Antibody (OTI2D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810280S
Description
MLH1 Monoclonal antibody specifically detects MLH1 in Human samples. It is validated for Western BlotSpecifications
| MLH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6PKH6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 317749 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D12 | |
| Unconjugated | |
| DHRS4L2 | |
| 1110035C23Rik; AI317206; AI325952; AI561766; COCA2; colon cancer, nonpolyposis type 2; DNA mismatch repair protein Mlh1; DNA mismatch repair protein Mlh1-like protein; FCC2; hMLH1; HNPCC; HNPCC2; I79_014127; MGC5172; mismatch repair protein 1; MLH1; MLH-1; mutL homolog 1; mutL homolog 1 (E. coli); mutL homolog 1, colon cancer, nonpolyposis type 2; mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli); mutL protein homolog 1 | |
| Full length human recombinant protein of human DHRS4L2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.