missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21066-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.Specifications
| MLKL | |
| Polyclonal | |
| Unconjugated | |
| MLKL | |
| BC028141, MLKL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 197259 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NB16 | |
| MLKL | |
| MLKL Fusion Protein Ag15188 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction