missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18824-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a small peptide hormone that is secreted by cells of the small intestine to regulate gastrointestinal contractions and motility. Proteolytic processing of the secreted protein produces the mature peptide and a byproduct referred to as motilin-associated peptide (MAP). Two transcript variants encoding different preproprotein isoforms but the same mature peptide have been found for this gene.Specifications
| MLN | |
| Polyclonal | |
| Unconjugated | |
| MLN | |
| MAP, MLN, motilin, Promotilin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4295 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12872 | |
| MLN | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction