Learn More
MLXIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13614-1-AP-20UL
 View more versions of this product
Product Code. 16821914
  • 145.00€ / 20µL
Description
This antibody could recognize isoform1 (∽110 kDa) and isoform3(∽69 kDa) of MLXIP. A prominent and specific MLXIP of approximately 130 kDa was detected. It demonstrates that MLXIP associated specifically with Mlx in the cytoplasm.
This gene encodes a protein that functions as part of a heterodimer to activate transcription. The encoded protein forms a heterodimer with Max-like protein X (MLX) and is involved in the regulation of genes in response to cellular glucose levels.
Specifications
Specifications
MLXIP
Polyclonal
Unconjugated
MLXIP
bHLHe36, KIAA0867, MIR, MLX interacting protein, MLXIP, MONDOA
Rabbit
Antigen Affinity Chromatography
RUO
208104, 22877, 304479
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2VPU4, Q9HAP2
Mlxip
MLXIP Fusion Protein Ag4519
20 μL
Primary
Human, Mouse, Rat