Learn More
MLXIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13614-1-AP-20UL
Description
This antibody could recognize isoform1 (∽110 kDa) and isoform3(∽69 kDa) of MLXIP. A prominent and specific MLXIP of approximately 130 kDa was detected. It demonstrates that MLXIP associated specifically with Mlx in the cytoplasm.
This gene encodes a protein that functions as part of a heterodimer to activate transcription. The encoded protein forms a heterodimer with Max-like protein X (MLX) and is involved in the regulation of genes in response to cellular glucose levels.Specifications
| MLXIP | |
| Polyclonal | |
| Unconjugated | |
| MLXIP | |
| bHLHe36, KIAA0867, MIR, MLX interacting protein, MLXIP, MONDOA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 208104, 22877, 304479 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q2VPU4, Q9HAP2 | |
| Mlxip | |
| MLXIP Fusion Protein Ag4519 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |