missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-7 Antibody (704202), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals FAB7170K
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MMP-7 Monoclonal antibody specifically detects MMP-7 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Especificaciones
| MMP-7 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 704202 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Mouse myeloma cell line NS0-derived recombinant mouse MMP-7, Met1-Leu264, Accession # Q10738 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway | |
| 4316 | |
| Store at 4°C in the dark. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido