missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€ - 544.00€
Specifications
| Antigen | MMP-9 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678187
|
Novus Biologicals
NBP2-41233-0.025mg |
0.025 mg |
189.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18156567
|
Novus Biologicals
NBP2-41233 |
0.1 mg |
544.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-9 Polyclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| MMP-9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| MMP9 | |
| IgG | |
| Affinity Purified | |
| 73 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| PBS with 0.02% Sodium Azide | |
| 4318 | |
| Antibody was raised against a 16 amino acid peptide near the center of human MMP9. The immunogen is located within amino acids 260 - 310 of MMP9 . Amino Acid Squence: PSERLYTQDGNADGK | |
| Primary | |
| At least three isoforms of MMP9 are known to exist; this antibody only recognizes the two longest isoforms. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title