Learn More
MMP-9 Antibody (CL0539) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52949APC
Product Code. 30298812
  • 737.00€ / 0.10mL
Description
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMP-9
Monoclonal
APC
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
This antibody was developed using a recombinant protein derived from P14780, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
4318
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0539
PBS
Mouse
Protein A purified