missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-9 Antibody (CL0539) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52949C
This item is not returnable.
View return policy
Description
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MMP-9 | |
| Monoclonal | |
| DyLight 650 | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| This antibody was developed using a recombinant protein derived from P14780, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| 4318 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0539 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction