Learn More
PeproTech MMP2 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P307-50UG
Description
AA Seq: MYNFFPRKPK WDKNQITYRI IGYTPDLDPE TVDDAFARAF QVWSDVTPLR FSRIHDGEAD IMINFGRWEH GDGYPFDGKD GLLAHAFAPG TGVGGDSHFD DDELWTLGEG QVVRVKYGNA DGEYCKFPFL FNGKEYNSCT DTGRSDGFLW CSTTYNFEKD GKYGFCPHEA LFTMGGNAEG QPCKFPFRFQ GTSYDSCTTE GRTDGYRWCG TTEDYDRDKK YGFCPETAMS TVGGNSEGAP CVFPFTFLGN KYESCTSAGR SDGKMWCATT ANYDDDRKWG FCPDQGYSLF LVAAHEFGHA MGLEHSQDPG ALMAPIYTYT KNFRLSQDDI KGIQELYGAS PDIDLGTGPT PTLGPVTPEI CKQDIVFDGI AQIRGEIFFF KDRFIWRTVT PRDKPMGPLL VATFWPELPE KIDAVYEAPQ EEKAVFFAGN EYWIYSASTL ERGYPKPLTS LGLPPDVQRV DAAFNWSKNK KTYIFAGDKF WRYNEVKKKM DPGFPKLIAD AWNAIPDNLD AVVDLQGGGH SYFFKGAYYL KLENQSLKSV KFGSIKSDWL GC Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MMP-2. Anti-Human MMP-2-specific antibody was purified by affinity chromatography employing an immobilized Human MMP-2 matrix. Sandwich ELISA: To detect Human MMP-2 by sandwich ELISA (using 100 μL/well) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MMP-2 (500-P307BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human MMP-2. Western Blot: To detect Human MMP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2μg/ml. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MMP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
MMP (matrix metalloproteinase) are proteolytic enzymes capable of degrading connective tissue components. MMP have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPa and TIMPs play a significant role in regulating angiogenesis. MMP2 is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids, and contains the basic structure of propeptide, catalytic, and hemopexin domains. The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane-bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non-fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc-binding site characterizes the structure of the MMPs. Functionally, MMP2 is involved in tissue remodeling. Mutations in MMP-2 gene have been associated with Winchester syndrome and Nodulosis-Arthropathy-Osteolysis (NAO) syndrome. Two transcript variants encoding different isoforms of MMP-2 have been found.
Specifications
| MMP2 | |
| Polyclonal | |
| PBS with no preservative | |
| P08253 | |
| MMP2 | |
| E.coli-derived, 62.0 kDa Recombinant Human MMP-2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| Unconjugated | |
| MMP2 | |
| 72 kDa gelatinase; 72 kDa type IV collagenase; 72 kDa type IV collagenase isoform a preproprotein; 72kD gelatinase; 72kD type IV collagenase; 72kDa gelatinase; 72kDa type IV collagenase; CLG 4A; CLG4; CLG4A; Collagenase; collagenase type IV-A; endopeptidase; GelA; gelatinase A; Gelatinase alpha; Mat; matrix metallo protease; matrix metallopeptidase 2; matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloprotease 2; matrix metalloproteinase; matrix metalloproteinase 2; matrix metalloproteinase 2 (72 KDa type IV collagenase); matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloproteinase-2; matrix metalloproteinase-II; MMP; MMP2; MMP-2; MMP-2 protein; MMP-II; MMPs; MONA; neutrophil gelatinase; PEX; prepro-72kDa matrix metalloproteinase; Progelatinase A; TBE1; TBE-1; wu:fa99h12; wu:fk89d01 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4313 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.