missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MMP28 293T Cell Transient Overexpression Lysate

Product Code. 18448789 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18448789 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18448789 Supplier Novus Biologicals™ Supplier No. H00079148T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Cancer
Target MMP28
Synonym EC 3.4.24.-, EPILYSIN, matrix metallopeptidase 28, matrix metalloproteinase 28, MM28, MMP25, MMP-28 MMP-25, putative MMP28 gene product
Gene Symbol MMP28
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.