missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MNAR Polyclonal Antibody

Rabbit Polyclonal Antibody
618.00€
Specifications
| Antigen | MNAR |
|---|---|
| Concentration | 0.05 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17139303
![]() |
Invitrogen™
PA5111384 |
100 μL |
618.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17179343
![]() |
Invitrogen™
PA5111429 |
100 μL |
618.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: GPPTTANHLG LSVPGLVSVP PRLLPGPENH RAGSNEDPIL APSGTPPPTI PPDETFGGRV PRPAFVHYDK EEASDVEISL ESDSDDSVVI VPE Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 94%.
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.Specifications
| MNAR | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| PELP1 | |
| 4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3 | |
| PELP1 | |
| Primary | |
| Antigen affinity chromatography |
| 0.05 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZL8 | |
| 27043 | |
| Recombinant Protein corresonding to Human MNAR. Recombinant protein control fragment (Product #RP-101664). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title