Learn More
Tuttnauer Mobile Lifting Device
Product Code. 17829548 Shop All Tuttnauer Products
Change view
Click to view available options
Product Type:
Lifting Device
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
17829548 Lifting Device Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17829548 Supplier Tuttnauer Supplier No. HLD5070044
  • ONLINE EXCLUSIVE PRICE
    11697.00€
    9189.30€ / Pack of 1
    Save 2507.70€
    21% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
Mobile lifting device for vertical autoclaves with heavy loads up to 100 kg. The lift is mobile, equipped with an integrated swivel arm for maximum maneuverability and can service a few autoclaves. Controlled by push buttons on a movable handset. Equipped with a maintenance free electrical battery.
  • The mobile lifting device for vertical autoclave basket with heavy sterilization load up to 100 kg.
  • The lift is mobile equipped with an integrated swivel arm for maximum maneuverability and can service a few autoclaves.
  • Controlled by push buttons on a movable control handset.
  • Equipped with a maintenance free electrical battery.
Specifications
Product Type Lifting Device