Learn More
Ansell Model 122 - AlphaTec™ 3000 Ultrasonically Welded and Taped Coverall
Product Code. 18842054 Shop All Ansell Products
Change view
Click to view available options
Size:
2X-Large
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
18842054 2X-Large Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18842054 Supplier Ansell Supplier No. YE30T0012206
  • ONLINE EXCLUSIVE PRICE
    737.10€
    465.03€ / Pack of 15
    Save 272.07€
    36% Off
In Stock
Add to basket
Add to basket
Durable, comfortable multi-layer chemical barrier, Type 3/4/5/6 protection.
  • One of the lightest and most comfortable chemical protective garments, this durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
  • Ultrasonically Welded And Taped Seams
  • Provides a strong liquid and particle barrier
  • 2 piece hood
  • Elasticated hood, waist, sleeve over cuffs and ankles
  • Double zip closure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Collared Coveralls
Size 2X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Double
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded, Taped
Sterility Non-sterile
Resistance Liquid, Anti-static, Particle, Bloodborne Pathogens