missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Molecular Biology Grade Water (DNase and RNase-free)

Product Code. 31908400 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31908400 100 mL 100mL
31908479 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31908400 Supplier Sartorius Supplier No. 018691B
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Molecular Biology Grade Water is suitable for cell-culture and molecular biology applications.

  • Undergoes reverse osmosis
  • Meets USP and EPTesting specifications
  • DNase and RNase-free
  • Suitable for cell-culture and molecular biology applications
  • Long-storage when handled properly under defined conditions
  • pH: 5.0 to 7.0
  • Appearance: Clear Solution
  • Endotoxins: test < 0.05 EU/mL
  • DNase detection: Not Detected
  • RNase detection: Not Detected

Specifications

Chemical Name or Material Water
Molecular Weight (g/mol) Unknown
Percent Purity Unknown
Grade Cell Culture/Molecular Biology
Packaging Bottle
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.