missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-IT™ LIVE Plasma Membrane and Nuclear Labeling Kit (Counterstains For GFP-Expressing Cells)

Product Code. 10778273
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10778273 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10778273 Supplier Invitrogen™ Supplier No. I34406
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Counterstains for GFP-expressing cells

The Image-iT LIVE Plasma Membrane and Nuclear Labeling Kit provides two stains - red-fluorescent Alexa Fluor 594 wheat germ agglutinin (WGA) and blue-fluorescent Hoechst 33342 dye - for highly selective staining of the plasma membrane and nucleus, respectively, of live green-fluorescent protein (GFP)-transfected cells.

Cell Analysis, Cell Structure, Cell-Based Reporter Assays, Cellular Imaging, Enzyme & Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Nucleus, Nucleoli & Nuclear Envelope, Plasma Membrane, Proteins, Expression, Isolation & Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red, Blue
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 1 kit
Detection Method Fluorescence
Product Line Image-iT
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Membrane and Nuclear Labeling
Sub Cellular Localization Nucleus, Plasma Membrane

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.