Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Blot Stain Kit
Description
The Pro-Q Diamond Phosphoprotein Blot Stain Kit provides a rapid and simple method for directly detecting phosphoproteins on polyvinylidene difluoride (PVDF) or nitrocellulose membranes. The fluorescent stain binds selectively to the phosphate moiety and binding is independent of the amino acid sequence context.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Detection Location | In-Blot Detection |
| Detection Method | Fluorescence |
| Label or Dye | Pro-Q Diamond |
| Quantity | 1 kit |
| Target Molecule | Proteins (Phosphoproteins) |
| Product Line | PRO-Q |
| Product Type | Phosphoprotein Blot Stain Kit |