missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum, TRC

Product Code. 30481792
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481792 10 g 10g
30473899 25 g 25g
30480151 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481792 Supplier Toronto Research Chemicals Supplier No. TRCM36100010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
Synonym Molybdenum, 3N5, Amperit 105.054, Amperit 106.2, MChVL, Metco 63, Molybdenum element, NSC 600660, NSC 600661, NSC 600665, NSC 603570, NSC 603571, NSC 603572, SGC 15, TMO 10, TMO 30, TMO 50, TMOIO, TsM1, Molybdaen, Molybdän
IUPAC Name Molybdenum
SMILES [Mo]

Specifications

Quantity 10 g
InChI Formula InChI=1S/Mo
Formula Weight 97.9054
Chemical Name or Material Molybdenum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.