Learn More
Monastrol, TRC
Product Code. 30483550
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483550 2.5 mg 2.5mg
30472836 10 mg 10mg
30475468 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30483550 Supplier Toronto Research Chemicals Supplier No. TRCM5057502.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 329689-23-8
Molecular Formula C14 H16 N2 O3 S
Molecular Weight (g/mol) 292.35
IUPAC Name ethyl 4-(3-hydroxyphenyl)-6-methyl-2-sulfanylidene-3,4-dihydro-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=C(C)NC(=S)NC1c2cccc(O)c2
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C14H16N2O3S/c1-3-19-13(18)11-8(2)15-14(20)16-12(11)9-5-4-6-10(17)7-9/h4-7,12,17H,3H2,1-2H3,(H2,15,16,20)
Formula Weight 292.0882
Chemical Name or Material Monastrol