missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus A29L Humanized Recombinant Human Monoclonal Antibody (P1D4)
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554750
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Monkeypox Virus A29L is a gene that is found in the genome of the Monkeypox virus (MPXV). MPXV is a member of the Orthopoxvirus genus, which also includes the variola virus (which causes smallpox) and the vaccinia virus (used in smallpox vaccination). The A29L gene encodes a protein that is involved in various aspects of the viral life cycle, including viral replication, assembly, and immune evasion.
Specifications
| Monkeypox Virus A29L Humanized | |
| Recombinant Monoclonal | |
| 5 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| VHH-hFc |
| ELISA | |
| P1D4 | |
| Unconjugated | |
| A29L | |
| Recombinant MPXV A29L. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction