missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus D8L (MPXV/USA/2022/MA001) Recombinant Rabbit Monoclonal Antibody (HL2950)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556798
This item is not returnable.
View return policy
Description
Monkeypox Virus D8L (MPXV/USA/2022/MA001) Recombinant Monoclonal Antibody for Western Blot
The Monkeypox Virus D8L gene encodes a protein that is integral to the structural integrity and infectivity of the monkeypox virus, which belongs to the Orthopoxvirus genus. The D8L protein, also known as the A-type inclusion (ATI) protein, is involved in the formation of A-type inclusion bodies within infected cells. These inclusion bodies are thought to play a role in protecting the virus from the host immune response and facilitating viral transmission. Monkeypox virus causes a zoonotic disease in humans that presents with symptoms similar to smallpox, including fever, rash, and lymphadenopathy, but is generally less severe. The D8L protein's role in the virus's life cycle and immune evasion makes it a significant target for research into antiviral therapies and vaccines. By studying the D8L protein, researchers aim to better understand the mechanisms of monkeypox virus pathogenesis and develop effective strategies to prevent and treat monkeypox infections, which have emerged as an increasing public health concern.
Specifications
| Monkeypox Virus D8L (MPXV/USA/2022/MA001) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2950 | |
| Unconjugated | |
| D8L | |
| Full length of Monkeypox virus D8L. (#MPXV_USA_2022_MA001). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction