missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus F13L (MPXV/USA/2022/MA001) Recombinant Rabbit Monoclonal Antibody (HL2637)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556799
This item is not returnable.
View return policy
Description
Monkeypox Virus F13L (MPXV/USA/2022/MA001) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The F13L gene of the Monkeypox virus encodes for a key protein involved in the formation and release of extracellular enveloped virions, which are crucial for the virus's ability to spread and infect host cells. This protein, also known as the p37 protein, is essential for the virus's life cycle and plays a significant role in the viral morphogenesis process. The F13L gene is highly conserved among orthopoxviruses, indicating its importance in viral replication and pathogenicity. Due to its critical role, the F13L protein is a potential target for antiviral drug development, aiming to inhibit the spread of the Monkeypox virus by disrupting the formation of infectious viral particles.
Specifications
| Monkeypox Virus F13L (MPXV/USA/2022/MA001) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2637 | |
| Unconjugated | |
| F13L | |
| Full length Monkeypox virus F13L protein. (MPXV_USA_2022_MA001). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction