missing translation for 'onlineSavingsMsg'
Learn More

Mono-2-ethylhexyl Phthalate Glucuronide, TRC

Product Code. 30009996
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009996 5 mg 5mg
30133264 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009996 Supplier Toronto Research Chemicals Supplier No. TRCM5425005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59348-65-1
Molecular Formula C22H30O10
Molecular Weight (g/mol) 454.47
Synonym 1-(2-Ethylhexyl 1,2-Benzenedicarboxylate) beta-D-Glucopyranuronic Acid
IUPAC Name (2S,3S,5R,6S)-6-[2-(2-ethylhexoxycarbonyl)benzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CCCCC(CC)COC(=O)c1ccccc1C(=O)O[C@@H]2O[C@@H]([C@@H](O)C(O)[C@H]2O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H30O10/c1-3-5-8-12(4-2)11-30-20(28)13-9-6-7-10-14(13)21(29)32-22-17(25)15(23)16(24)18(31-22)19(26)27/h6-7,9-10,12,15-18,22-25H,3-5,8,11H2,1-2H3,(H,26,27)/t12?,15?,16-,17+,18-,22-/m0/s1
Formula Weight 454.1839
Chemical Name or Material Mono-2-ethylhexyl Phthalate Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.