missing translation for 'onlineSavingsMsg'
Learn More

Mono[2-(perfluorohexyl)ethyl] phosphate, TRC

Product Code. 30007137
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007137 100 mg 100mg
30006891 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007137 Supplier Toronto Research Chemicals Supplier No. TRCM566800100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57678-01-0
Molecular Formula C8 H6 F13 O4 P
Molecular Weight (g/mol) 444.08
Synonym 6:2 monoPAP, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-(dihydrogen phosphate)-1-octanol
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl dihydrogen phosphate
SMILES OP(=O)(O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H6F13O4P/c9-3(10,1-2-25-26(22,23)24)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h1-2H2,(H2,22,23,24)
Formula Weight 443.9796
Chemical Name or Material Mono[2-(perfluorohexyl)ethyl] phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.