missing translation for 'onlineSavingsMsg'
Learn More

Mono N-Desmethyl Disperse Polyester Dark Blue, TRC

Product Code. 30012465
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012465 10 mg 10mg
30009600 1 mg 1mg
30004802 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012465 Supplier Toronto Research Chemicals Supplier No. TRCM53949510MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56524-77-7
Molecular Formula C15 H12 N2 O4
Molecular Weight (g/mol) 284.27
Synonym 9,10-Anthracenedione, 1-amino-4,5-dihydroxy-8-(methylamino)-
IUPAC Name 1-amino-4,5-dihydroxy-8-(methylamino)anthracene-9,10-dione
SMILES CNc1ccc(O)c2C(=O)c3c(O)ccc(N)c3C(=O)c12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H12N2O4/c1-17-7-3-5-9(19)13-11(7)14(20)10-6(16)2-4-8(18)12(10)15(13)21/h2-5,17-19H,16H2,1H3
Formula Weight 284.0797
Chemical Name or Material 1-Amino-4,5-dihydroxy-8-(methylamino)anthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.