missing translation for 'onlineSavingsMsg'
Learn More

Mono(n-pentyl) Terephthalate, TRC

Product Code. 30482774
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482774 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482774 Supplier Toronto Research Chemicals Supplier No. TRCM566603250MG
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mono(n-pentyl) Terephthalate
CAS 1819-30-3
Molecular Formula C13 H16 O4
Quantity 250 mg
Synonym Terephthalic Acid Monopentyl Ester, Terephthalic Acid Pentyl Ester, p-(Pentyloxycarbonyl)benzoic Acid
InChI Formula InChI=1S/C13H16O4/c1-2-3-4-9-17-13(16)11-7-5-10(6-8-11)12(14)15/h5-8H,2-4,9H2,1H3,(H,14,15)
SMILES CCCCCOC(=O)c1ccc(cc1)C(=O)O
IUPAC Name 4-pentoxycarbonylbenzoic acid
Molecular Weight (g/mol) 236.264
Formula Weight 236.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.