missing translation for 'onlineSavingsMsg'
Learn More

Mono-tert-butyl Malonate, TRC

Product Code. 30478031
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478031 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478031 Supplier Toronto Research Chemicals Supplier No. TRCM52503010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mono-tert-butyl Malonate
CAS 40052-13-9
Molecular Formula C7H12O4
Quantity 10 g
Synonym Propanedioic Acid 1-(1,1-Dimethylethyl) Ester, Propanedioic Acid Mono(1,1-dimethylethyl) Ester, 2-(tert-Butoxycarbonyl)acetic Acid, 3-tert-Butoxy-3-oxopropanoic Acid, Malonic Acid Mono-tert-butyl Ester, Malonic Acid tert-Butyl Monoester, tert-Butyl Hydrogen Malonate
InChI Formula InChI=1S/C7H12O4/c1-7(2,3)11-6(10)4-5(8)9/h4H2,1-3H3,(H,8,9)
SMILES CC(C)(C)OC(=O)CC(=O)O
IUPAC Name 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
Molecular Weight (g/mol) 160.17
Formula Weight 160.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.