missing translation for 'onlineSavingsMsg'
Learn More

Monobenzyl Phthalate Beta-D-Glucuronide, TRC

Product Code. 30138161
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138161 50 mg 50mg
30011817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138161 Supplier Toronto Research Chemicals Supplier No. TRCM52491050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102674-29-3
Molecular Formula C21H20O10
Molecular Weight (g/mol) 432.38
Synonym β-D-Glucopyranuronic acid 1-[2-(Phenylmethyl) 1,2-Benzenedicarboxylate]
SMILES O=C(O[C@@H]([C@@H]1O)O[C@H](C(O)=O)[C@@H](O)[C@@H]1O)C2=CC=CC=C2C(OCC3=CC=CC=C3)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H20O10/c22-14-15(23)17(18(25)26)30-21(16(14)24)31-20(28)13-9-5-4-8-12(13)19(27)29-10-11-6-2-1-3-7-11/h1-9,14-17,21-24H,10H2,(H,25,26)/t14-,15-,16+,17-,21-/m0/s1
Formula Weight 432.1056
Chemical Name or Material Monobenzyl Phthalate beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.