Learn More
Monobutyl Phthalate Acyl-Beta-D-glucuronide, TRC
Product Code. 30003829
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003829 5 mg 5mg
30009773 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30003829 Supplier Toronto Research Chemicals Supplier No. TRCM5251055MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85209-81-0
Molecular Formula C18H22O10
Molecular Weight (g/mol) 398.36
Synonym 1-(Butyl 1,2-Benzenedicarboxylate)-β-D-glucopyranuronic Acid, Monobutyl Phthalate Glucuronide;
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C18H22O10/c1-2-3-8-26-16(24)9-6-4-5-7-10(9)17(25)28-18-13(21)11(19)12(20)14(27-18)15(22)23/h4-7,11-14,18-21H,2-3,8H2,1H3,(H,22,23)/t11-,12-,13+,14-,18-/m0/s1
Chemical Name or Material Monobutyl Phthalate Acyl-beta-D-glucuronide