missing translation for 'onlineSavingsMsg'
Learn More

Monodesmethylchlorpheniramine, TRC

Product Code. 30134770
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134770 25 mg 25mg
30138250 50 mg 50mg
30010559 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134770 Supplier Toronto Research Chemicals Supplier No. TRCM53952025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20619-12-9
Molecular Formula C15H17ClN2
Molecular Weight (g/mol) 260.76
Synonym gamma-(4-Chlorophenyl)-N-methyl-2-pyridinepropanamine;Monodemethylchlorpheniramine, Chlorpheniramine EP Impurity C;
IUPAC Name 3-(4-chlorophenyl)-N-methyl-3-pyridin-2-ylpropan-1-amine
SMILES CNCCC(c1ccc(Cl)cc1)c2ccccn2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H17ClN2/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12/h2-8,10,14,17H,9,11H2,1H3
Formula Weight 260.108
Chemical Name or Material Monodesmethylchlorpheniramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.