missing translation for 'onlineSavingsMsg'
Learn More

Monomethylarsonous Acid, TRC

Product Code. 30479797
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479797 10 mg 10mg
30465153 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479797 Supplier Toronto Research Chemicals Supplier No. TRCM56510010MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25400-23-1
Molecular Formula CH5AsO2
Molecular Weight (g/mol) 123.97
Synonym Methanearsonous Acid, Methylarsonous Acid
IUPAC Name methylarsonous acid
SMILES O[As](O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/CH5AsO2/c1-2(3)4/h3-4H,1H3
Formula Weight 123.9506
Percent Purity >95%
Chemical Name or Material Monomethylarsonous Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.