missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monopolin Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-131F
This item is not returnable.
View return policy
Description
Monopolin Polyclonal antibody specifically detects Monopolin in Yeast samples. It is validated for Western Blot
Specifications
| Monopolin | |
| Polyclonal | |
| Western Blot | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Yeast | |
| Purified |
| Western Blot | |
| FITC | |
| PBS | |
| Synthetic peptide corresponding to a portion of human Monopolin. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction